Would you like to log in again?
Eisco® Connecting Leads with Alligator Clips
Supplier: EISCO SCIENTIFIC LLC
Eisco®
PH1053BPK12
470345-242EA 75.85 USD InStock
470345-242
Eisco® Connecting Leads with Alligator Clips
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Red connecting leads, each 300 mm in length with alligator clips on both ends for 4 mm plug.
  • 12" length (300 mm)
  • Alligator clips at both ends
  • Insulated
  • Quick, temporary electrical connections
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty