Would you like to log in again?
Heavy Duty Multi-Purpose Table Clamp
Supplier: EISCO SCIENTIFIC LLC
CH0688FBG
470345-290EA 26.2 USD InStock
470345-290
Heavy Duty Multi-Purpose Table Clamp
Clamps General Purpose Clamps
Made of die-cast metal, this clamp features a vinyl-coated grip and swivel pad, along with a built-in rod and pulley holder for versatile applications.
  • High performance, heavy duty multi-purpose table clamp
  • It's versatile design allows you to secure a wide variety of laboratory apparatus and accessories
  • Non-ferrous, drop forged steel construction that will not bend or break under heavy loads
  • Convenient built-in rod or pulley holder accommodates rods and pulleys up to 16 mm
  • Features a vinyl sleeve and swivel pad to ensure a strong, slip-proof grip on a variety of surfaces
  • Designed for use in professional laboratories, but is an ideal choice for science classrooms
Can be used for holding rods and pulleys, immobilizing meter sticks, securing lab equipment, and can also function as a G-Clamp. Made from heavy duty, die-cast metal, and features a vinyl sleeve and swivel pad to ensure a strong, slip proof grip on a variety of surfaces, up to 65 mm (2.5") thick. Non-ferrous, drop forged steel construction that will not bend or break under heavy loads. Solid metal screw adjustments and handles won't break while tightening. The built-in holder can accommodate rods or pulleys measuring up to 16 mm in diameter. Clamp measures approximately 5" wide, 4"tall and 1.5" long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty