Would you like to log in again?
Eisco Semi-Plano Glass Lenses
Supplier: Eisco Scientific
Eisco
PH0532DCV PH0532ICC PH0532CCV PH0532KCC PH0532JCC PH0532FCV PH0532ECV PH0532LCC
470350-616EA 5.1 USD InStock
470350-616 470350-624 470350-622 470350-620 470344-792 470345-300 470344-038 470350-618
Eisco Semi-Plano Glass Lenses
Educational Materials Physics Educational Materials Optics Learning Activities
Individual round glass lenses made with one flat side.
  • Plano-concave or plano-convex lenses available in a variety of focal lengths
  • 50 mm (2") Ø
  • Made of clear, optically-worked glass
  • Highly polished surface, ground edges
These plano-convex and plano-concave lenses feature one flat side and one curved side - common in the real world, but unusual for physics classrooms.
Eisco labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control.
Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments. These lenses are designed for educational use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty