Would you like to log in again?
Eisco Glass Filter Flasks, Pack of 2
Supplier: EISCO SCIENTIFIC LLC
Eisco
FL2402 FL2403 FL2404
470344-932EA 29.85 USD InStock
470344-932 470345-094 470345-356
Eisco Glass Filter Flasks, Pack of 2
Flasks Filter Flasks
Pack of two filter flasks with heavy walls and integral side arm made of borosilicate glass.
  • Withstands high temperatures and thermal shocks
  • Features tubulated integral side arm adaptable to ³/₈" internal diameter tubing
  • Heavy wall design for safe use under vacuum pressure
  • Optimized mouth and neck ensures secure fit with stopper when under pressure (stopper not included)
Pack of two filtering flasks with superior chemical durability and heat tolerance. The tubulated integral side arm, heavy walls, and enforced mouth and neck are all designed to safely withstand vacuum pressurization. Made of high quality, heavy duty 3.3 borosilicate glass with the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty