Would you like to log in again?
Eisco Bunsen Burner, LPG / Butane Gas - Heavy Base, Stopcock
Supplier: EISCO SCIENTIFIC LLC
CH0086B
470345-362EA 33.95 USD InStock
470345-362
Eisco Bunsen Burner, LPG / Butane Gas - Heavy Base, Stopcock
Heaters Gas Burners
Gas control stopcock to regulate and switch off gas flow.
  • Suitable for use with LPG (liquid petroleum gas) or butane gas
  • Serrated inlet arm accepts 6 to 8 mm tubing
  • Heavy casted base for additional stability
  • Nickel plated brass burner tube features a gas control stopcock to regulate and switch off the gas flow
Premium quality Bunsen burner, useful for a variety of laboratory applications and experiments, as well as glasswork and smoldering. Burner tube size: 8 mm Ø × 0.75" long. Overall size: 6.5" tall × 3" base Ø.
Caution: ⚠ WARNING
Cancer and Reproductive Harm - www.p65warnings.ca.gov
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty