Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Pipette Filler
Supplier: EISCO SCIENTIFIC LLC
CH0580D
470345-384EA 16.7 USD InStock
470345-384
Pipette Filler
Pipettes Pipette Bulbs
Silicon bulb pipette filler with a 54 mm diameter and three pinch valves, suitable for handling noxious solutions and providing precise and controlled liquid dispensing.
  • Eisco Labs Silicon bulb pipette filler
  • For pipetting noxious solutions, microbial suspensions and other harmful and unpleasant solutions
  • Comprises a bulb and three pinch valves controlling influx and efflux of pipette contents
  • Bulb diameter is 54 mm
Silicone bulb pipette filler, for pipetting noxious solutions, microbial suspensions and other harmful solutions. Comprises a bulb and three pinch valves controlling influx and efflux of pipette contents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty