Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Eisco® Vernier Calliper
Supplier: EISCO SCIENTIFIC LLC
Eisco®
PH0078A
470345-388EA 7.29 USD InStock
470345-388
Eisco® Vernier Calliper
Calipers
Precise and accurate, measures up to 12 cm with 0.1 mm graduations, made from high-quality materials, perfect for use in workshops and classrooms.
  • Dual scale: 0 - 12 cm×0.1 mm, 0 - 5"×1/16"
  • Inside, outside, or depth measurement
EISCO Vernier plated steel caliper for measuring inside, outside, or depth. Thumb wheel provided for easy motion of jaws.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty