Would you like to log in again?
Eisco Glass Volumetric Flasks with Glass Stopper, ASTM Class A
Supplier: EISCO SCIENTIFIC LLC
CH0441AWT CH0441BWT CH0441EWT CH0441GWT CH0441HWT CH0441H CH0441CWT CH0441DWT CH0441A02 CH0441D CH0441FWT CH0441A02WT CH0441A01
470343-958EA 6.35 USD InStock
470343-958 470345-604 470344-210 470344-994 470344-696 470344-626 470344-744 470346-050 470343-942 470345-420 470345-992 470345-126 470345-114
Eisco Glass Volumetric Flasks with Glass Stopper, ASTM Class A
Flasks Volumetric Flasks
ASTM Class A volumetric flasks are made from borosilicate 3.3 glass and feature a graduation mark, solid glass pennyhead stopper, and blue or white enamel markings.
  • Made of high quality, heavy duty, borosilicate 3.3 glass for excellent durability and chemical resistance
  • Class A tolerances conform with ASTM E288 standards for accuracy
  • Fitted with a solid glass pennyhead stopper
  • Available with blue or white enamel markings and specifications
Calibrated to contain.
Eisco labs borosilicate glass has the following heat tolerances:
- Strain point 515 °C
- Annealing point 565 °C
- Softening point 820 °C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty