Would you like to log in again?
Hardwood Double Burette Stand
Supplier: EISCO SCIENTIFIC LLC
CH0264B
470345-494EA 32.3 USD InStock
470345-494
Hardwood Double Burette Stand
Support Stands
Double burette stand crafted from seasoned hardwood.
  • Burette stand with sturdy seasoned hardwood construction will stand up to wear and tear in the lab
  • Stand can accomodate two burettes up to 10 mm in diameter.
  • Wooden base (300×100 mm) with threaded hole, wooden support dowel (460×16 mm), and double sided clamp with three nylon thumb screws included
  • Designed for use in professional laboratories, also an ideal choice for science classrooms
Double burette rack made of seasoned hardwood stands 19.5" tall with double arm to hold a burette on each side. Smallest allowable diameter burette or tube is 10 mm diameter. Base measures 298.45×107.95 mm (11.75x4.25") with a threaded hole for the wooden support dowel. Black nylon thumb screws. Flasks and burette in picture not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty