Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco ASTM Glass Graduated Cylinders, Class A

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CH0341B CH0341HBL CH0341E CH0341D CH0341G CH0341A05BL CH0341F CH0341EBL
470345-326EA 75.6 USD InStock
470345-326 470345-502 470345-548 470343-636 470345-716 470345-340 470344-188 470344-510
Eisco ASTM Glass Graduated Cylinders, Class A
Cylinders Measuring Cylinders
The highest quality and precision in cylinders.

  • Made of high quality, heavy duty, borosilicate 3.3 glass
  • Conforms with ASTM E1272 Class A tolerance standards
  • Hexagonal base provides maximum stability and prevents rolling when placed on side
  • Easy-to-read graduations and specifications
  • Fitted with a removable, plastic protective collar

Spout for easy pouring, and hexagonal base for extra stability. Class A tolerance conforms with ISO/ASTM. Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks. Calibrated to contain.

Eisco Labs Borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C

Packaging: One unit per pack.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty