Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Eisco Support Rings with Stem, Steel
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0668B CH0668D CH0668C CH0668A
470344-112EA 4.49 USD InStock
470344-112 470345-284 470344-176 470345-522
Eisco Support Rings with Stem, Steel
Support Rings
Closed metal support rings with long stems great for supporting funnels.
  • Closed retort support ring with stem made from plated mild steel
  • Supports standard or separatory funnel, or use with triangles to support flat bottomed glassware
  • Stem diameter measures 0.8 cm (0.3"), stem length 14 cm
Use with bosshead clamp (available separately) to attach to a support rod.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty