Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Glass Volumetric Flasks with PE Stopper, Class A, Includes Calibration Certificate

Supplier: EISCO SCIENTIFIC LLC
Eisco
CH1952J CH1952E CH1952H CH1952I CH1952G CH1952F CH1952E01 CH1952E02 CH1952D
470345-722EA 9.15 USD InStock
470345-722 470343-592 470345-728 470345-606 470344-160 470345-480 470345-670 470344-434 470344-532
Eisco Glass Volumetric Flasks with PE Stopper, Class A, Includes Calibration Certificate
Flasks Volumetric Flasks
Class A borosilicate glass Volumetric flasks with PE stoppers and calibration certificates.

  • Withstands high temperatures and thermal shocks
  • Single blue graduation mark
  • Printed with unique alpha numeric serial
  • Individually calibrated at 20 °C

Volumetric flask made of high quality, heavy duty Borosilicate 3.3 glass. Calibrated to contain. Eisco borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C

Certifications: Class A tolerances as per DIN ISO 1042 standards.

Delivery information: Includes an interchangeable 10/19 polypropylene stopper and calibration certificate.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty