Would you like to log in again?
32-Peg Draining Rack
Supplier: EISCO SCIENTIFIC LLC
CH0715B
470345-712EA 140.8 USD InStock
470345-712
32-Peg Draining Rack
Racks Draining Racks
This metal rack features 32 pegs for efficient drying of labware, ensuring proper airflow and organization in the laboratory.
  • Test tube draining rack featuring a sturdy vinyl-coated steel construction that will stand up to the wear and tear of the lab
  • Supports up to 32 test tubes with 4.25" pegs - Provides convenient insertion and removal of tubes. 21.5" tall, 17.25" wide, 12" depth from the peg tips, and weighs 2.6 lbs.
  • Stands upright or hangs on the wall (mounting hardware not included)
  • Designed for use in professional laboratories, but is an ideal choice for science classrooms
Vinyl coated metal build is easy on glassware while being durable at the same time. Pegs are spaced 2.25" apart horizontally and 6/5.5/6.5" vertically to hold a variety of shapes and sizes; test tubes, beakers, flasks and more.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty