Would you like to log in again?
Hoffman Electrolysis Apparatus
Supplier: EISCO SCIENTIFIC LLC
CH0740B
470345-734EA 163.69 USD InStock
470345-734
Hoffman Electrolysis Apparatus
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
This apparatus, made of borosilicate glass, features PTFE stopcocks and includes platinum and carbon electrodes, providing a comprehensive setup for electrolysis.
  • Used for demonstrating the electrolytic decomposition of water or determining chemical composition
  • Made from heavy duty borosilicate 3.3 glass with glass key stopcock
The Hoffman Electrolysis Apparatus is used for the demonstration of the chemical composition of water and the production of gases through chemical reactions. The borosilicate glass unit consists of two 50 ml capacity outer tubes graduated in 0.2 ml increments, connected to a central tube with a 200 ml-capacity funnel-shaped bulb. The supplied platinum electrodes mounted in rubber stoppers are used for the electrolysis of acidified water; carbon electrodes are supplied for ammonia solutions, hydrochloric acid, or other chloride-containing solutions. Includes an instruction guide.
Ordering information: A support stand with clamps (model CH0740F) and 12 V DC power supply are required but NOT included.
Delivery information: Includes hoffman electrolysis apparatus with two PTFE stopcocks, two carbon electrodes in rubber stoppers, two platinum electrodes in rubber stoppers, and instruction guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty