Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Glass Filter Funnels - Heavy

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Eisco
CH0458C CH0458D
470345-810EA 10.8 USD InStock
470345-810 470344-588
Eisco Glass Filter Funnels - Heavy
Funnels Filtration Funnels Filter Funnels
Heavy, thick-walled filter funnels with 60° angles.

  • Made of high quality Borosilicate 3.3 glass that is built to last and withstand multiple uses
  • Heavy construction provides greater mechanical strength and prolonged life with harder than normal usage
  • Features thick, uniform walls that allow for increased durability, ease of use and continual flow of liquid

Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.

Eisco Labs Borosilicate glass has the following heat tolerances:
- Strain point 515 °C
- Annealing point 565 °C
- Softening point 820 °C

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty