Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Porcelain Crucible with Lid, Tall Form

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CH0074G CH0074A10 CH0074B CH0074D CH0074E CH0074F CH0074F10
470345-108EA 4.29 USD InStock
470345-108 470344-612 470344-372 470344-574 470343-732 470345-398 470345-830
Eisco Porcelain Crucible with Lid, Tall Form
Crucibles
Can withstand temperatures up to 1900 °F (1050 °C).

  • Crucible for basic laboratory use to heat chemical compounds
  • Tall form body with lid
  • Constructed of glazed porcelain for strength, thermal performance, and enhanced chemical resistance

Tall form high-quality glazed porcelain crucibles with lids. Great for heating chemical compounds in the lab.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty