Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Glass Separatory Funnels with Glass Stopcock

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Eisco
CH0482C CH0482E CH0482D CH0482B CH0482F CH0482A
470345-832EA 21 USD InStock
470345-832 470345-414 470343-932 470344-034 470345-234 470343-684
Eisco Glass Separatory Funnels with Glass Stopcock
Funnels Separating Funnels
Separatory funnels with ground cones, plastic stoppers, and glass key stopcocks, made of borosilicate 3.3 glass.

  • Made of high quality, heavy-duty borosilicate 3.3 glass - can withstand high temperatures and thermal shocks
  • Used to separate the components of a mixture into two immiscible liquids of different densities
  • Fitted with a glass key stopcock for precise drop-wise dispensing of the denser separated liquid
  • Features a ground glass cone with and interchangeable plastic stopper
  • Ungraduated

Ungraduated, pear-shaped separating funnels are made of high quality borosilicate glass. They feature a ground cone, plastic stopper, and glass key stopcock. Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.

Eisco Labs borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty