Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Glass Graduated Cylinders with Stopper and Round Base

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CH0348C CH0348H CH0348E CH0348G CH0348B CH0348A CH0348F CH0348D
470345-612EA 28.3 USD InStock
470345-612 470344-844 470345-856 470344-722 470344-130 470345-472 470343-568 470344-730
Eisco Glass Graduated Cylinders with Stopper and Round Base
Cylinders Measuring Cylinders
High-quality borosilicate Class B glass cylinders with an interchangeable polypropylene stopper and round base.

  • Made of high quality, heavy duty, borosilicate 3.3 glass
  • Conforms with DIN ISO 4788 Class B tolerance standards
  • No spout
  • Easy-to-read blue graduations and specifications contrast against light color liquids

Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks. Calibrated to contain.

Eisco labs borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C

Packaging: One unit per pack.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty