Would you like to log in again?
Swivel Clamp Holder
Supplier: EISCO SCIENTIFIC LLC
CH0688E
470345-904EA 29.75 USD InStock
470345-904
Swivel Clamp Holder
Clamps General Purpose Clamps
With the ability to tilt clamps at any angle, this clamp holder accommodates large rods up to 21 mm and offers convenient screw adjustability.
  • High performance, jumbo sized, swivel clamp holder made of die-casted metal
  • The holder allows you to tilt clamps at any angle in parallel planes
  • Screw adjustable clamp accommodates rods of up to 21 mm in diameter
  • Designed for use in professional laboratories, but is an ideal choice for science classrooms
The clamp holder measures 7.25" in length.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty