Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Glass Powder Funnels - Jointed

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Eisco
CH0460C CH0460F CH0460E CH0460A CH0460B CH0460D
470345-490EA 13.5 USD InStock
470345-490 470344-726 470345-916 470344-594 470346-016 470344-256
Eisco Glass Powder Funnels - Jointed
Funnels Powder Funnels
Jointed powder funnels with ground cones and 60° angles.

  • Made of high quality borosilicate 3.3 glass that is built to last and withstand multiple uses
  • Features a ground cone for greater seal and extra stability when jointed to glassware
  • Durable, uniform walls allow for ease of use and continual flow of liquid

Low temperature gradient - can withstand high temperatures and thermal shocks.

Eisco Labs Borosilicate glass has the following heat tolerances:
- Strain point 515 °C
- Annealing point 565 °C
- Softening point 820 °C

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty