Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gas Preparation Kit

Supplier: EISCO SCIENTIFIC LLC
CH0889
470345-960EA 667.99 USD InStock
470345-960
Gas Preparation Kit
Labware Sets
This 26-piece set includes all the essential glass elements for gas production, providing convenience and reliability in gas preparation processes, all made of durable borosilicate glass for optimal performance and chemical resistance.

  • Made of high quality, heavy duty 3.3 borosilicate glass
  • 26 piece gas preparation kit
  • Contains all essential glass elements for quick setup for production of gasses
  • Supplied in two gratnells trays, with each glassware piece securely tucked into fitted foam packaging
  • Perfect for any classroom or laboratory

This kit contains all essential glass elements needed for quick setup for production of gases in lab including Ammonia, Bromine, Carbon Dioxide, Carbon Monoxide, Chlorine, Hydrogen, Hydrogen Bromide, Hydrogen Iodide, Hydrogen Chloride, Hydrogen Sulphide, Hydrogen Peroxide, Nitric Oxide, Nitrogen, Nitrogen Dioxide, Nitrous Oxide, Oxygen, Sulphur Dioxide, Sulphur Trioxide etc.

Complete kit is supplied in 2 Gratnells trays, each glassware piece is secured in a neatly cut foam packaging. The kit consists of the following components:
Recovery bend 24/29
Bend tube
Adjustable wash bottle head 24/29
T adapter
Test tube 24/19
Combustion tube
Dropping funnel 50 cm³
Gas mixing tube
Test tube with side arm
2x cone/screw cap adapter
Delivery adapter 24/29
2x gas cover jars 75 mm diameter
Dropping funnel plug 10/19
Dreshel bottle 125 cm³ 24/29
Beehive shelf 75 mm diameter 24/29
Flask R.B. 150 cm³ 24/29
5x Glass tuber 7 mm o.d.
Plastic trough
2x Gas jar 200×50 mm
Instruction manual

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty