Would you like to log in again?
Eisco Glass Conical Measures, Ungraduated
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0355G CH0355F
470345-622EA 28.5 USD InStock
470345-622 470345-966
Eisco Glass Conical Measures, Ungraduated
Cylinders Measuring Cylinders
Ungraduate conical measure made of borosilicate glass, with pouring spout and round base.
  • Premium glass conical measuring cup made of durable borosilicate glass
  • Ungraduated
  • Pouring spout makes for convenient and accurate distribution of liquids
  • Comes with a stable yet elegant rounded base
  • Autoclavable and dishwasher safe
Borosilicate 3.3 Glass conical measuring cup has a pouring spout for ease of distributing liquids in the lab. Wide extra stable piece. Ungraduated. Autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures & thermal shocks.
Eisco Labs Borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty