Would you like to log in again?
Eisco Support Rings with Bosshead Clamp, Cast Iron
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0674A CH0674D CH0674E CH0674C CH0674B
470344-934EA 9.29 USD InStock
470344-934 470345-286 470344-820 470344-126 470345-984
Eisco Support Rings with Bosshead Clamp, Cast Iron
Support Rings
Heavy closed cast iron support rings with integral bosshead clamps great for supporting funnels.
  • Closed retort support ring with integral bosshead made from zinc-plated cast iron
  • Supports standard or separatory funnels, or use with a triangle to support flat bottomed glassware
  • Distance from rod attachment to center of ring measures 12 cm (4.7"), allowing for vertical alignment across different diameter rings
  • Bosshead fits 8 to 1.6 cm (0.6") diameter rods
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty