Would you like to log in again?
Eisco® Single-Sided Hardwood Metric Meter Stick with Vertical Reading and Zero Top
Supplier: EISCO SCIENTIFIC LLC
Eisco®
PH0064CPK10 PH0064C
470345-990EA 8.29 USD InStock
470345-990 470345-202
Eisco® Single-Sided Hardwood Metric Meter Stick with Vertical Reading and Zero Top
Rulers
This single-sided hardwood meter stick with vertical reading and a zero top is ideal for classroom or lab experiments requiring precise measurements.
  • Durable hardwood meter stick with zero top and vertical reading
  • Overall dimensions: 100×2.5 cm (6 mm thickness)
  • Great for the classroom
Measurements are graduated in centimeters and millimeters. Also available in packs of 10.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty