Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Glass Volumetric Flasks with PE Stopper, Class A, USP, Includes Calibration Certificate

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Eisco
CH1972E CH1972C CH1972H CH1972G CH1972B CH1972J CH1972D CH1972I
470343-658EA 18.7 USD InStock
470343-658 470344-284 470346-010 470343-998 470345-908 470344-708 470343-842 470345-698
Eisco Glass Volumetric Flasks with PE Stopper, Class A, USP, Includes Calibration Certificate
Flasks Volumetric Flasks
Class A USP borosilicate glass Volumetric flasks with PE stoppers and calibration certificates.

  • Withstands high temperatures and thermal shocks
  • Single blue graduation mark
  • Printed with unique alpha numeric serial
  • Individually calibrated at 20 °C

Volumetric flask made of high quality, heavy duty Borosilicate 3.3 glass. Calibrated to contain. Eisco borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C

Certifications: Conforms with USP 31 standards for food, drug, or medicinal use. Class A, conforms with DIN ISO 1042 standards.

Delivery information: Includes an interchangeable 10/19 polypropylene stopper and calibration certificate.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty