Would you like to log in again?
Eisco Glass Gilson Separatory Funnels with Glass Stopcock
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0487C CH0487E CH0487D CH0487B CH0487A
470345-250EA 34.6 USD InStock
470345-250 470344-662 470343-972 470345-188 470346-028
Eisco Glass Gilson Separatory Funnels with Glass Stopcock
Funnels Separating Funnels
Ungraduated Gilson separatory funnels with ground cones, plastic stoppers, and glass key stopcocks, made of borosilicate 3.3 glass.
  • Made of high quality, heavy-duty borosilicate 3.3 glass - can withstand high temperatures and thermal shocks
  • Used to separate the components of a mixture into two immiscible liquids of different densities
  • Gilson style features a long delivery stem for good control of the liquid/liquid interface
  • Fitted with a glass key stopcock for precise drop-wise dispensing of the denser separated liquid
  • With ground glass cone with and interchangeable plastic stopper
  • Ungraduated
Gilson style Separating Funnels have a long delivery stem for good control of the liquid/liquid interface. They feature a ground cone, plastic stopper, and glass key stopcock. Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.
Eisco Labs borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty