Would you like to log in again?
Eisco® High Resolution, Spring Balances with Dual Scale
Supplier: EISCO SCIENTIFIC LLC
Eisco®
PH0024C PH0024E PH0024B PH0024A PH0024D
470346-014EA 4.89 USD InStock
470346-014 470344-948 470346-042 470344-996 470345-368
Eisco® High Resolution, Spring Balances with Dual Scale
Balances and Scales
High-resolution spring balance with transparent dual scale in Newtons and grams. Zero calibration capability and acrylic body for durability.
  • Premium quality spring balances
  • Acrylic body with dual transparent scale - measures in Newtons and grams
  • User friendly zero-point calibration capability
  • Ideal for physics demonstrations and experiments
  • Spring balance scale measures 9" tall in total, 1.4" wide and 0.85" long
User friendly zero-point calibration capability- simply pull up or push down the tab at the top of the scale to adjust. Features a loop at the top for hanging and a hook at the base to hold items being measured. Ideal for physics classrooms, demonstrations and experiments. Height of the scale body is 5.4".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty