Would you like to log in again?
Eisco Lens Holder for Optics Bench
Supplier: EISCO SCIENTIFIC LLC
Eisco
PH0652C
470346-066EA 9 USD InStock
470346-066
Eisco Lens Holder for Optics Bench
Educational Materials Physics Educational Materials Optics Learning Activities
Lens holder for 50 mm lenses or mirrors for use with Eisco Optical Bench.
  • Plastic lens holder for lenses or mirrors with a 50 mm diameter
  • Can be secured to a base or bench with attached 8 mm grooved rod
  • Great for use with optical benches
Plastic Lens Holder accommodates lenses or mirrors with a 50 mm diameter. Holder can be attached to a base or bench using the 8 mm diameter rod with grooves on the bottom of the lens holder.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty