Would you like to log in again?
Eisco Glass Büchner Funnels with Plain Stem
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0466B CH0466C CH0466E CH0466A CH0466D
470344-636EA 13.1 USD InStock
470344-636 470345-658 470345-082 470346-084 470345-078
Eisco Glass Büchner Funnels with Plain Stem
Funnels Filtration Funnels Büchner Funnels
Büchner funnels with G3 porosity sintered discs made of borosilicate 3.3 glass.
  • Made of high-quality borosilicate 3.3 glass that is built to last and withstand multiple uses
  • Features a G3 porosity sintered disc
  • Straight, thick, uniform walls for increased durability, ease of use, and continual flow of liquid
Borosilicate glass Büchner funnels with straight sides and G3 porosity sintered discs. Eisco Labs Borosilicate glass has the following heat tolerances: Strain point 515 °C; Annealing point 565 °C; Softening point 820 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty