Would you like to log in again?
Eisco® Push Pull Spring Scales
Supplier: EISCO SCIENTIFIC LLC
Eisco®
PH0035D PH0035E PH0035A PH0035C PH0035G PH0035B
470345-348EA 10.59 USD InStock
470345-348 470346-114 470346-054 470344-688 470345-574 470344-840
Eisco® Push Pull Spring Scales
Balances and Scales
Acrylic push/pull balances with different capacities. Can be used to measure both pushing and pulling forces. Ideal for use in classroom for physics experiments and measuring weights of small objects.
  • Transparent acrylic tubular body
  • Spring mechanism is clearly visible
  • Calibrated dual scale
The blue push pull balance (250 g/2.5 N) is designed for measuring force, others allow you to use grams for a mass activity and Newtons for a force activity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty