Would you like to log in again?
UniStore™ Amber Reagent Bottles, Narrow Mouth, HDPE
Supplier: United Scientific Supplies
UniStore™
33429 33427 33428 33426 33425 33422 33421 33424 33423
470346-134EA 24.2 USD InStock
470346-134 470346-198 470346-142 470346-130 470346-196 470346-150 470346-182 470346-190 470346-126
UniStore™ Amber Reagent Bottles, Narrow Mouth, HDPE
Bottles Plastic Bottles
Leakproof, amber laboratory quality bottles with many applications in the field or laboratory.
  • Protect light-sensitive materials during shipping and storage
  • Bottles meet USP requirements for storage of light-sensitive materials
  • Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact
  • Free of heavy metals, with leakproof closure featuring an integrated seal ring
  • Precision molded for consistent wall thickness and smooth interior and exterior surfaces
UniStore™ narrow mouth HDPE reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant. The amber bottles are designed to reduce UV light transmission to protect light-sensitive contents, and conform to USP requirements for storage of light-sensitive materials. Excellent chemical resistance to most acids, bases, and alcohols.
Ordering information: Polypropylene caps are included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty