Would you like to log in again?
Motic Swift Line M3-F-BTI1 Comparison LED Microscope with Tablet
Supplier: MOTIC INSTRUMENTS USA, INC SE
Motic
M3-F-BTI1
470346-262EA 3186.65 USD InStock
470346-262
Motic Swift Line M3-F-BTI1 Comparison LED Microscope with Tablet
Microscopes
A truly unique concept in microscopy!
  • 8" LCD Android tablet with 1280×800 screen resolution
  • Connection: WiFi; micro SD card
  • WiFi (2.4 or 5 GHz) and Ethernet (RJ45)
  • MotiConnect App
  • Connect five WiFi devices at one time
The M3-F forensic microscope offers professional features and detachable 8" tablet at a highly competitive price point. Use the M3-F's revolutionary technology to compare images in both micro and macro environments. The M3's dedicated macro lens, with a large working distance, allows you to view not only bullets but also larger more bulky, 'evidence' items.
Swift's powerful optical system allows for images to be seen either 100% from the left microscope, 100% from the right, side-by-side, or overlapping. The M3-F is the first and only comparison microscope at this price level that can achieve overlapping of images.
Combining the features of Swift's popular M3-F's advanced comparison microscope with an Adroid based tablet system offers unlimited possibilities.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty