Would you like to log in again?
Motic Swift Line M17T Advanced Compound Microscopes with Tablet
Supplier: MOTIC INSTRUMENTS USA, INC SE
Motic
M17T-BTI1-P M17T-BTI2-P
470346-266EA 2632.4 USD InStock
470346-266 470346-264
Motic Swift Line M17T Advanced Compound Microscopes with Tablet
Microscopes
The latest in Swift Optical's line of digital compound microscopes featuring integrated LCD tablets.
  • All-in-one digital microscope
  • MotiConnect app: image-capture, annotation, measurement and sharing functions
  • 4×, 10×, 40× and 100× objectives
The M17 compound microscope series features infinity corrected plan optics, ideal for college laboratories, veterinary and medical applications. Supplied with trinocular head, WiFi camera with bracket, and 8 or 10" tablet, these research level microscopes also incorporate 3 W LED illumination and wide field 20 mm eyepieces for a very crisp image.
Motic Swift Line’s BTI Series represents the latest in educational technology. Capable of sharing images using its advanced WiFi system, this Android-based tablet includes live image viewing app, MotiConnect. The BTI Series feature high-speed, full resolution imaging technology built into one of our most popular microscopes. Use it as a conventional microscope or share live images with your class using WiFi tablets and wireless laptops. Connect up to five devices.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty