Would you like to log in again?
Motic Swift Line M30TZ-BTI Zoom Stereo Microscope with Tablet
Supplier: MOTIC INSTRUMENTS USA, INC SE
Motic
M30TZSM99CLBTI
470346-268EA 2028.5 USD InStock
470346-268
Motic Swift Line M30TZ-BTI Zoom Stereo Microscope with Tablet
Microscopes
The latest in Swift Optical's line of digital Stereo microscopes featuring integrated LCD Tablets.
  • Paired widefield eyepieces are 45° inclined, with diopter adjustment. Interpupillary adjustment from 55 to 75 mm
  • Interlocked eyepiece tubes with diopter for increased focus acuity
  • Viewing head rotates 360°, locks in place by tightening thumb screw
The M30TZ with Tablet is ideal for laboratory, industrial and educational use. This stereo microscope features zoom objectives with a continuous magnification range of 7.5× to 40×, offering added versatility that is not an option with fixed power stereo microscopes.
Motic Swift Line’s BTI Series represents the latest in educational technology. Capable of sharing images using its advanced WiFi system, this Android-based tablet includes live image viewing app, MotiConnect.
The BTI Series feature high-speed, full resolution imaging technology built into one of our most popular microscopes. Use it as a conventional microscope or share live images with your class using WiFi tablets and wireless laptops. Connect up to five devices.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty