Would you like to log in again?
Eisco® Bell in Vacuum Jar
Supplier: EISCO SCIENTIFIC LLC
Eisco®
PH0176CN8
470346-452EA 73.45 USD InStock
470346-452
Eisco® Bell in Vacuum Jar
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Bell in vacuum jar is a classic experiment demonstrating properties of sound.
  • Demonstrate the dependence of sound on the medium through which it travels
  • Bell jar made of high-quality, break-resistant acrylic
  • Electric bell operaties on 4 to 6V DC power source (not included)
This experiment demonstrates that sound needs a material medium for transmission, and it helps to reinforce the concept that sound is a mechanical wave that requires particles to carry its energy. Slowly decrease the air inside with a vacuum pump (available separately) and hear the sound disappear. Electric bell operates on 4 to 6V DC power supply. Size of bell: 25 cm tall×14 cm diameter.
Ordering information: Requires a vacuum pump and DC power supply (not included).
Delivery information: Includes 1 bell jar, 1 electric bell, 1 rubber cork with terminals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty