Would you like to log in again?
Eisco Laser Ray Box
Supplier: EISCO SCIENTIFIC LLC
Eisco
LRBX
470346-456EA 156.3 USD InStock
470346-456
Eisco Laser Ray Box
Educational Materials Physics Educational Materials Optics Learning Activities
This laser ray box is adjustable, magnetic, and bright enough to use in ambient light.
  • Operates on 5 V USB power supply or rechargeable battery
  • Produces either 3 or 5 parallel laser beams
  • Bright enough to use in a classroom setting with ambient light
  • Magnetic strip included for whiteboard use
When the traditional white light ray boxes do not provide strong enough rays to be seen across a classroom of ambient light, then we recommend this 635 nm wavelength, 3/5 ray, monochromatic (laser) ray box. The battery powered ray box comes with a USB cable so it can easily be charged with a computer, a charging station, or a standard USB wall adapter. An included magnetic strip may be applied to the back of the ray box so that it can be used with a metalic white board for demonstrations. Easily toggle between three or five parallel laser rays.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty