Would you like to log in again?
Steris Amsco 3080 Surgical Table
Supplier: GUMBO MEDICAL
S3080ST
470346-652EA 11818.19 USD InStock
470346-652
Steris Amsco 3080 Surgical Table
Furniture Tables and Workbenches
A surgical standard.
  • Refurbished
  • 1 year parts warranty
  • Includes new mattress
  • For educational and training use only
The Steris®/Amsco® General Surgical Table is one of the most reliable and popular surgical tables. Trusted for years by healthcare facilities around the world for its proven performance, the 3080 is a cost-effective solution for general Operating Rooms and training centers.
The 3080 familiar, easy to use hand control quickly performs Bieri hydraulic ultra-precise tabletop articulations for any patient posture needed. Self-compensating hydraulic floor locks adjust automatically for uneven surfaces, firmly fixing the table in place. The table includes a return to level feature, enhancing patient safety. T-shaped pedestal design and reverse orientation provide optimal access to the patient by the entire surgical team, regardless of procedure. The 3080 also has a streamlined radiolucent tabletop for easy C-arm access and complete upper and lower body image amplification (IA) coverage. An optional Radiolucent Orthopedic Extension can quickly convert this workhorse general surgical table into a fracture table. In addition, Bariatric Width Extenders stretch the table width to 28" to accommodate the girth of up to a 500 lbs. (227 kg) patient.
The 3080 RL/SP handles virtually all general surgical specialty procedures such as Pediatrics, Urology, Neurology, Bariatrics, ENT, OB/GYN, Endovascular, Plastic Surgery, Ophthalmology, and Orthopedics (including skeletal traction, with accessories).
Delivery information: This item has to ship by freight, Additional shipping charges may apply.
Caution: This product is refurbished and is intended for educational and training use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty