Would you like to log in again?
Laser Optical Demonstration Set
Supplier: AMERICAN SCIENTIFIC LLC SE
5133-00
470346-740EA 142.65 USD InStock
470346-740
Laser Optical Demonstration Set
Educational Materials Physics Educational Materials Optics Learning Activities
This kit uses a compact yet powerful laser ray box, which has bright and well-defined rays perfect for classroom use.
  • Includes teacher's guide
  • Concave and convex lens
  • Parallel flat or parallel bar
  • Trapezoid prism
  • Right angle lens, mirror
  • Hollow semicircle and frosted semicircle
  • Laser light beam chart
This kit uses a compact yet powerful laser ray box, which has bright and well-defined rays perfect for classroom use. The laser ray box projects 1, 3, or 5 parallel laser beams onto any flat surface ideal for optical ray viewing and analysis. It includes everything needed to demonstrate the basic principles of geometrical optics. A convenient, impact-resistant, foam-lined black carrying case keeps everything organized and ready to use. The magnetic activity mat measures 381L×210W mm (15×8.25") with printed scales and angles.
Delivery information: The Ray Box is powered by an AC adapter (included).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty