Would you like to log in again?
Eisco® DIY Miniature Catapult
Supplier: EISCO SCIENTIFIC LLC
Eisco®
GP00007MINI
470346-872EA 12.6 USD InStock
470346-872
Eisco® DIY Miniature Catapult
Educational Materials Physics Educational Materials Mechanics Learning Activities
Miniature catapult for demonstrating STEM concepts with a medieval weapon.
  • Easy to build and fun to use
  • Miniature 3.5" length is perfect for desktops and side tables, or for individual student building
  • Introduces and demonstrates real world STEM concepts
  • Includes instructions for assembly and experimental activities
Catapults were one of the most influential elements of siege warfare over the last 2500 years. The easy to build design allows students, teachers, or science lovers to play with the physics of catapults. The small size makes the catapult excellent for individual building, and makes an interesting desktop or side table piece. Educational topics this kit can be used to demonstrate includes force, acceleration, center-of-mass, vector components, Hooke’s law, parabolic trajectories, and class 2 levers.
Kit includes catapult base, catapult left and right sides (symmetric except for counter-sinks), catapult lever arm, 8 rubber bands, 4 wood screws, 3 dowels, 1 c-shaped screw hook, 1 ping pong ball, cardboard storage box, and instructions for assembly and experimental activities. (A medium size Phillips screwdriver is required but not included). Catalpult is approximately 3.5" long when assembled.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty