Would you like to log in again?
Light Emitting Diodes
Supplier: United Scientific Supplies
RDJMBLEDPK10 YLWLEDPK10 YELJMBLEDPK10 GRNJMBLEDPK10 ORGLEDPK10 RDLEDPK5 GNLEDPK5
470346-950EA 6.69 USD InStock
470346-950 470346-948 470346-958 470346-966 470346-944 470346-932 470346-964
Light Emitting Diodes
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Quality LEDs for electrical experiments.
  • 2 pin connectors
  • Fits all standard bred boards
  • Available in two sizes
These LEDs fit perfectly into a Bred board for investigating electrical circuits. By their nature, these LEDs will only work correctly in DC circuits with the proper electrical orientation. Each LED has a Cathode (– shorter side) and Anode (+ longer side).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty