Would you like to log in again?
Eisco® Ring and Strings for Force Table
Supplier: EISCO SCIENTIFIC LLC
CENCO®
PH0347CN8RINGS
470346-968EA 12.35 USD InStock
470346-968
Eisco® Ring and Strings for Force Table
Educational Materials Physics Educational Materials
Replacement ring and strings for force table.
  • Consists of one top ring and four 12" strings with terminal rings for attaching masses
Place center ring over central hub of table and drape radial strings over four pulleys attached 90 degrees from each other. Replacement part for Force Table part number 470014-356.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty