Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Welch Allyn® DS 66 Palm Aneroids
Supplier: Healthcare Products
Welch Allyn®
WEL5098-27CB WEL5098-30CB WEL5098-28CB WEL5098-29CB
470347-150EA 591.29 USD InStock
470347-150 470347-072 470347-060 470347-018
Welch Allyn® DS 66 Palm Aneroids
Blood Pressure Monitors
Easy to use and read.
  • Soft comfort grip
  • Jewel movement for long life
  • Laser-engraved dial face for accuracy
  • Trigger deflation valve for smooth, one-finger operation
Maintain accurate blood pressure readings with lower lifetime cost with gear-free DuraShock™ technology, the first major innovation in aneroid technology in nearly 100 years. Part of the Gold Series, the DS66 Trigger features an innovative trigger-release design for easy operation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty