Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Welch Allyn® CP150 Compact Resting ECG
Supplier: Healthcare Products
Welch Allyn®
WEL105410 WEL714730 WEL720705 WEL719653 WEL105660 WELCP150AWD-1ENB WEL410370 WELCP150AS-1ENB WEL105353 WELCP150A-1ENB WEL105343 WELCP150AW-1ENB
470347-210EA 1096.17 USD InStock
470347-210 470347-110 470347-176 470347-168 470347-194 470347-064 470347-118 470347-280 470347-268 470347-114 470347-238 470347-248
Welch Allyn® CP150 Compact Resting ECG
Data Loggers
Simple and easy to use design.
  • Durable and high-quality materials
  • 7" TPT Color-touch screen interface
  • Rechargeable battery can last up to 8 hours of continuous operation
The Welch Allyn® CP 150 Compact Resting Electrocardiograph is a simple and efficient 12-Lead EKG machine aimed to help medical professionals in diagnosing and monitoring heart conditions by recording the electrical activity of the heart at rest. This machine can provide information about heart rate and rhythm, and can detect problems such as heart enlargement, history of heart attack (myocardial infarction) and other conditions, which can greatly help in properly treating the patient.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty