Would you like to log in again?
Baxter® ELI 380 Resting ECG
Supplier: Healthcare Products
Baxter®
WELELI380-DCS11 WELELI380-ACX11
470347-252EA 39834.96 USD InStock
470347-252 470347-258
Baxter® ELI 380 Resting ECG
Data Loggers
Reduce the need for repeat ECGs.
  • Streamlined barcode workflow eliminates steps and reduces manual entry
  • Glass surfaces on the capacitive touchscreen display simplify cleaning
  • Follows ACC, AHA and HRS recommendations for baseline filtering (0.05 Hz)
  • Exceeds ACC, AHA and HRS upper frequency cutoff (300 Hz) for adults and children
  • DoD-approved security features safeguard patient health information (PHI) and your network
  • Critical Test Results help quickly identify high-risk patients
  • 12-lead or 15-lead ECG
The ELI380 Resting ECG, with ERGO swivel and touchscreen display, is designed to make the total ECG experience more efficient-from decreasing the time required to perform the procedure, to increasing the accuracy and security of the data acquired, to the overall patient care experience.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty