Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Welch Allyn® Two-Part Reusable Blood Pressure Cuffs
Supplier: Healthcare Products
Welch Allyn®
WEL5082-42 WEL5082-26 WEL5082-44 WEL5089-21 WEL5082-11 WEL5082-16 WEL5089-02 WEL5082-01H WEL5089-22 WEL5082-25 WEL5089-01H WEL5089-25 WEL5082-24 WEL5082-01 WEL5082-03 WEL5082-02 WEL5089-03 WEL5089-22H WEL5089-04 WEL5082-07 WEL5082-08 WEL5089-01 WEL5082-43
470347-166EA 102.12 USD InStock
470347-166 470347-234 470347-112 470347-024 470347-162 470347-272 470347-084 470347-260 470347-120 470347-230 470347-010 470347-098 470347-130 470346-978 470347-090 470346-996 470346-982 470347-206 470347-228 470347-038 470347-302 470347-216 470347-314
Welch Allyn® Two-Part Reusable Blood Pressure Cuffs
Blood Pressure Monitors
Replace that worn out cuff.
  • One or two tube options
  • Multiple sizes
  • Replacement cuff and bladder options
The Welch Allyn® two-piece reusable blood pressure cuff is a traditional style two-piece blood pressure cuff with removable cuff and bladder. It is designed for use with large aneroid, wall aneroid, hand aneroid, and mercury sphygmomanometers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty