Would you like to log in again?
Compound Research Microscope, Swift Optical Instruments
Supplier: MOTIC INSTRUMENTS USA, INC SE
MAX-812PL
470347-386EA 1282.05 USD InStock
470347-386
Compound Research Microscope, Swift Optical Instruments
Microscopes
With a wide range of features for university and laboratory use, this advanced research microscope features binocular head, Koehler illumination, and infinity corrected plan optics.
  • Inward-facing quintuple revolving nosepiece
  • 3W LED Koehler illuminator with intensity control
  • Built-in low-drive mechanical stage (160×140 mm) with extended post for X and Y control
  • Abbe/Iris condenser with slot for contrast slider
  • 20 mm eyepieces with rubber eyeshields
  • 30 degree inclined ergonomic viewing head
Internal 110 to 220 V auto-switching power supply is built-in. Ergonomically designed, professional frame with low-position coaxial focusing allows one-handed operation, making extended use comfortable.
Corrected Infinity Optical System, EC Infinity Plan Achromatic 4×/0.10 (WD 7.18 mm), 10×/0.25 (WD 4.8 mm), 40×/0.65 (WD 0.68 mm), 100×/1.25/S-Oil (WD 0.2 mm).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty