Would you like to log in again?
Ecology Under the Microscope Microslide
Supplier: SUPERTEK SCIENTIFIC LLC
T098-15 T-98
470009-072PK 12.7 USD InStock
470009-072 470347-428
Ecology Under the Microscope Microslide
Slides Prepared Slides
Investigate ecological processes and life cycles.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson plan on the topic under study
Old atoms never die. Atoms that may have once belonged to a dinosaur may now be in your blood and tissues. This set shows students how the microscopic world helps to keep the world ecosystem in balance as it follows the on-going cycle of producers, consumers and decomposers.
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable Microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Contents:
Algae – Diatom (200×) / Pediastrum (200×)
Protozoa – Blepharisma (200×) / Suctorian (200×)
Starfish embryos (200×)
Arthropods – Cyclops (10×)
Nematodes (40X)
Virus on E. coli (60000×)
Fungi – Penicillium (200×)
Bacteria – Spirillum (400×)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty