Would you like to log in again?
Plant Tissue Macronutrients Test Kit
Supplier: Aldon
IS9758
470347-438EA 142.75 USD InStock
470347-438
Plant Tissue Macronutrients Test Kit
Environmental Test Kits
Easy way to test plant samples for their required macronutrients.
  • 50 tests for macronutrient presence in plant tissue samples
  • Aligned with GLOBE protocols
Plants need a variety of nutrients and minerals to grow and maintain overall health. The 3 main nutrients that plants require to survive and thrive are nitrogen, phosphorus, and potassium (also known as NPK). In this activity, students will extract nutrients from plants of choice and perform analyses to determine if nitrogen, phosphorus, and potassium concentration is abundant, adequate, or deficient.
Delivery information: Includes 500 ml nutrient extractor solution; 30 ml nitrogen reagent #1; 30 g nitrate reagent # 2; 30 g phosphorous reagent #1; 30 ml phosphorous reagent #2; 30 g potassium reagent #1; 120 ml potassium reagent #2; 100 pack filter paper; 4 collection bottles; 1 spot plate; 2 reagent scoops; 1 stirring rod; 5 pipettes.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty