Would you like to log in again?
Innovating Science® Phenol Red pH Indicator Kit
Supplier: ALDON CORP SE
Innovating Science®
IS5043
470347-442EA 10.15 USD InStock
470347-442
Innovating Science® Phenol Red pH Indicator Kit
Environmental Test Kits
Easy way to test water for pH within range 6.8 to 8.2.
  • Over 50 tests to test pH of water samples
Test water pH within 6.8 to 8.2 range with this phenol red pH testing kit.
Delivery information: Includes 30 ml phenol red, 0.02%; 2 test tube; 2 pipettes; 1 phenol red pH color chart.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty