Would you like to log in again?
Urban Water Test Kit
Supplier: Aldon
IS9762
470347-444EA 132.55 USD InStock
470347-444
Urban Water Test Kit
Environmental Test Kits
Easy way to test urban water for multiple factors.
  • 10 tests for urban water samples
Test water sample for chloride, copper, iron, hardness, nitrate, pH, phosphates, and temperature.
Delivery information: Includes 3 water collection bottles; 6 reaction vessesls; 6 measuring tubes; 15 pipettes; 6×1 ml titrators; 2×3 ml titrators; 1 pack toothpicks; 10 phosphate powder pillows; 1 pack pH strips; 10 iron test tablets; 10 copper test tablets; 5 g coliform test powder; 3×100 ml silver nitrate titrant; 3×15 ml salinity indicator solution; 3×60 ml EDTA titrant; 60 ml pH 10 buffer solution; 15 ml Erichrome black T solution; 60 ml Griess reagent; 0.5 g cadmium metal powder.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty